missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2,5-dichloropyridine 98.0+%, TCI America™

Catalog No. A24441G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24441G Supplier TCI America Supplier No. A24441G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78607-32-6
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.001
MDL Number MFCD00204157
InChI Key OLZPJUVEGSNIJL-UHFFFAOYSA-N
Synonym 3-amino-2,5-dichloropyridine, 2,5-dichloro-3-aminopyridine, 2,5-dichloro-pyridin-3-ylamine, 3-pyridinamine, 2,5-dichloro, 2,5-dichloro-3-pyridylamine, 2,5-dichloropyridine-3-amine, pubchem5507, acmc-209per, ksc495k8j
PubChem CID 2799999
IUPAC Name 2,5-dichloropyridin-3-amine
SMILES C1=C(C(=NC=C1Cl)Cl)N

Specifications

Melting Point 127°C
Quantity 1 g
Formula Weight 163.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2,5-dichloropyridine
TSCA No
Recommended Storage Refrigerator