accessibility menu, dialog, popup

UserName

NH2HNONH2

CAS RN 785-30-8

IUPAC Name: 4-amino-N-(4-aminophenyl)benzamide
Synonyms: 4-amino-N-(4-aminophenyl)benzamide
Molecular Weight (g/mol): 227.27
Molecular Formula: C13H13N3O
InChi Key: XPAQFJJCWGSXGJ-UHFFFAOYSA-N
SMILES: NC1=CC=C(NC(=O)C2=CC=C(N)C=C2)C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)