accessibility menu, dialog, popup

UserName

H3CNONHOCH3ClNHN

CAS RN 783348-36-7

IUPAC Name: N-{6-chloro-7-methoxy-9H-pyrido[3,4-b]indol-8-yl}-2-methylpyridine-3-carboxamide
Synonyms: N-{6-chloro-7-methoxy-9H-pyrido[3,4-b]indol-8-yl}-2-methylpyridine-3-carboxamide
Molecular Weight (g/mol): 366.81
Molecular Formula: C19H15ClN4O2
InChi Key: ZNOLRTPMNMPLHY-UHFFFAOYSA-N
SMILES: COC1=C(NC(=O)C2=C(C)N=CC=C2)C2=C(C=C1Cl)C1=C(N2)C=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results