accessibility menu, dialog, popup

UserName

CAS RN 78111-17-8

CH3(R)(S)OO(S)H3C(S)HO(S)O(R)(R)(R)O(R)H3C(S)(R)O(S)(R)HO(R)H3COHOHOOCH3O(R)OHH2C(E)

CAS RN 78111-17-8

IUPAC Name: (2R)-3-[(2S,5R,6R,8S)-8-[(2R,3E)-4-[(2R,4'aR,5R,6'S,8'R)-8'-hydroxy-6'-[(1S,3S)-1-hydroxy-3-[(3R,6S)-3-methyl-1,7-dioxaspiro[5.5]undecan-2-yl]butyl]-7'-methylidene-hexahydro-3'H-spiro[oxolane-2,2'-pyrano[3,2-b]pyran]-5-yl]but-3-en-2-yl]-5-hydroxy-10-methyl-1,7-dioxaspiro[5.5]undec-10-en-2-yl]-2-hydroxy-2-methylpropanoic acid
Synonyms: okadaic acid
Molecular Weight (g/mol): 805.02
Molecular Formula: C44H68O13
InChi Key: QNDVLZJODHBUFM-AAWJMCDUSA-N
SMILES: C[C@@H](C[C@H](O)[C@H]1O[C@@H]2CC[C@@]3(CC[C@@H](O3)\C=C\[C@@H](C)[C@@H]3CC(C)=C[C@@]4(O[C@H](C[C@@](C)(O)C(O)=O)CC[C@H]4O)O3)OC2[C@H](O)C1=C)C1O[C@@]2(CCCCO2)CC[C@H]1C
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results