missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Okadaic Acid, Prorocentrum sp.

Catalog No. 495604500UG
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
495604500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 495604500UG Supplier MilliporeSigma™ Supplier No. 495604500UG

Okadaic Acid, CAS 78111-17-8, is a highly potent inhibitor of protein phosphatase 1 (IC50 = 10-15 nM) and 2A (IC50 = 0.1 nM).

Packaged Under Inert Gas

Specifications

Color Colorless
Target Proteins Protein phosphatase
CAS 78111-17-8
Molecular Formula C44H68O13
Formula Weight 805g/mol
Solubility MeOH, EtOH, DMF, CHCl3
Content And Storage Protect from light
Form Glassy Solid
Quantity 500 μg
Purity 95% by HPLC