accessibility menu, dialog, popup

UserName

NH4OHSOOO

CAS RN 7803-63-6

IUPAC Name: ammonium hydrogen sulfate
Synonyms: ammonium bisulfate
Molecular Weight (g/mol): 115.1
Molecular Formula: H5NO4S
InChi Key: BIGPRXCJEDHCLP-UHFFFAOYSA-N
SMILES: [NH4+].OS([O-])(=O)=O
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (3)
Physical Form 
  • (1)
  • (2)
  • (2)
  • (3)
Grade 
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (7)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)