missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Bisulfate, Crystal, Reagent, 98%, Spectrum™ Chemical

Catalog No. 18600570
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600570 500 g Amber Glass Bottle
18600569 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18600570 Supplier Spectrum Chemical Mfg Cor Supplier No. A1140500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A1140, 7803-63-6, NH4HSO4

Spectrum™ Chemical Ammonium Bisulfate, Crystal, Reagent , also known as ammonium hydrogen sulfate, can be used as an alternative to sulfuric acid. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 7803-63-6
Molecular Formula H5NO4S
Molecular Weight (g/mol) 115.10
InChI Key BIGPRXCJEDHCLP-UHFFFAOYSA-N
IUPAC Name ammonium hydrogen sulfate
SMILES [NH4+].OS([O-])(=O)=O

Specifications

CAS Max % 100%
Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
UN Number UN2506
Percent Purity 98%
Grade Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Ammonium Bisulfate, Crystal