accessibility menu, dialog, popup

UserName

CH3H3CBr

CAS RN 78-77-3

IUPAC Name: 1-bromo-2-methylpropane
Synonyms: isobutyl bromide
Molecular Weight (g/mol): 137.02
Molecular Formula: C4H9Br
InChi Key: HLVFKOKELQSXIQ-UHFFFAOYSA-N
SMILES: CC(C)CBr
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)