accessibility menu, dialog, popup

UserName

CH3OOPOOOCH3OOCH3

CAS RN 78-51-3

IUPAC Name: tris(2-butoxyethyl) phosphate
Synonyms: tris(2-butoxyethyl) phosphate tris(butoxyethyl)phosphate tri(2-butoxyethanol)phosphate kronitex KP-140 phosflex T-bep tris-(2-butoxyethyl)fosfat TBEP tributyl cellosolve phosphate
Molecular Weight (g/mol): 398.48
Molecular Formula: C18H39O7P
InChi Key: WTLBZVNBAKMVDP-UHFFFAOYSA-N
SMILES: CCCCOCCOP(=O)(OCCOCCCC)OCCOCCCC
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)