missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-butoxyethyl) Phosphate 95.0+%, TCI America™

Catalog No. P0683500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0683500G 500 g
P068325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0683500G Supplier TCI America Supplier No. P0683500G
Only null left

Chemical Identifiers

CAS 78-51-3
Molecular Formula C18H39O7P
Molecular Weight (g/mol) 398.48
MDL Number MFCD00009456
InChI Key WTLBZVNBAKMVDP-UHFFFAOYSA-N
Synonym tris 2-butoxyethyl phosphate, tbep, phosflex t-bep, tris butoxyethyl phosphate, tributoxyethyl phosphate, tri butoxyethyl phosphate, kronitex kp-140, tributyl cellosolve phosphate, ethanol, 2-butoxy-, phosphate 3:1, tri 2-butoxyethyl phosphate
PubChem CID 6540
ChEBI CHEBI:35038
IUPAC Name tris(2-butoxyethyl) phosphate
SMILES CCCCOCCOP(=O)(OCCOCCCC)OCCOCCCC

Specifications

Color Yellow
Boiling Point 220°C
Quantity 500 g
Formula Weight 398.48
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(2-butoxyethyl) Phosphate
EINECSNumber (2)-2022
RTECSNumber KJ9800000
TSCA Yes