accessibility menu, dialog, popup

UserName

CAS RN 78-36-4

CH3OOH3CH3CH3CCH2

CAS RN 78-36-4

IUPAC Name: 3,7-dimethylocta-1,6-dien-3-yl butanoate
Synonyms: linalyl butyrate
Molecular Weight (g/mol): 224.34
Molecular Formula: C14H24O2
InChi Key: FHLGUOHLUFIAAA-UHFFFAOYNA-N
SMILES: CCCC(=O)OC(C)(CCC=C(C)C)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results