missing translation for 'onlineSavingsMsg'
Learn More

Linalyl Butyrate 97.0+%, TCI America™

Catalog No. L02755ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02755ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L02755ML Supplier TCI America Supplier No. L02755ML
Only null left

Specifications

CAS 78-36-4
Color Yellow
Boiling Point 82°C
Molecular Formula C14H24O2
Quantity 5 mL
MDL Number MFCD00048716
Synonym Butyric Acid Linalyl Ester
InChI Key FHLGUOHLUFIAAA-UHFFFAOYNA-N
SMILES CCCC(=O)OC(C)(CCC=C(C)C)C=C
IUPAC Name 3,7-dimethylocta-1,6-dien-3-yl butanoate
Molecular Weight (g/mol) 224.34
PubChem CID 62321
Formula Weight 224.34
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Linalyl Butyrate
Show More Show Less
EINECSNumber (2)-0763
TSCA Yes