accessibility menu, dialog, popup

UserName

CAS RN 77987-49-6

HONHOO

CAS RN 77987-49-6

IUPAC Name: benzyl N-(2-hydroxyethyl)carbamate
Synonyms: benzyl N-(2-hydroxyethyl)carbamate
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: SAGINAGERRNGGV-UHFFFAOYSA-N
SMILES: OCCNC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)