missing translation for 'onlineSavingsMsg'
Learn More

2-(Carbobenzoxyamino)-1-ethanol 97.0+%, TCI America™

Catalog No. H07935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07935G 5 g
H079325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07935G Supplier TCI America Supplier No. H07935G

Chemical Identifiers

CAS 77987-49-6
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00191060
InChI Key SAGINAGERRNGGV-UHFFFAOYSA-N
Synonym benzyl n-2-hydroxyethyl carbamate, z-glycinol, benzyl 2-hydroxyethyl carbamate, n-z-ethanolamine, 2-cbz-amino-1-ethanol, benzyl 2-hydroxyethylcarbamate, 2-z-amino ethanol, n-2-hydroxyethyl phenylmethoxy carboxamide, n-2-hydroxyethyl carbamic acid benzyl ester
PubChem CID 280458
IUPAC Name benzyl N-(2-hydroxyethyl)carbamate
SMILES OCCNC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 60°C
Boiling Point 215°C
Color White
Quantity 5 g
Formula Weight 195.22
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Carbobenzoxyamino)-1-ethanol