CAS RN 7791-11-9
CAS RN 7791-11-9
IUPAC Name:
rubidium(1+) chloride
Synonyms:
rubidium chloride
Molecular Weight (g/mol):
120.92
Molecular Formula:
ClRb
InChi Key:
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
SMILES:
[Cl-].[Rb+]
1
–
1
of
1
results
1
Rubidium chloride, 99.99%, (trace metal basis), Thermo Scientific™
CAS: 7791-11-9 Molecular Formula: ClRb Molecular Weight (g/mol): 120.92 InChI Key: FGDZQCVHDSGLHJ-UHFFFAOYSA-M Synonym: rubidium chloride,rubidium chloride rbcl,rubidium monochloride,rbcl,rubidium 1+ chloride,rubidium 1+ ion chloride,unii-n3shc5273s,rubidiumchloride,rubidium-chlorid,rubidium-chloride PubChem CID: 62683 ChEBI: CHEBI:78672 IUPAC Name: rubidium(1+);chloride SMILES: [Cl-].[Rb+]