accessibility menu, dialog, popup

UserName

CAS RN 7790-79-6

FFCd

CAS RN 7790-79-6

IUPAC Name: cadmium(2+) difluoride
Synonyms: cadmium fluoride
Molecular Weight (g/mol): 150.41
Molecular Formula: CdF2
InChi Key: LVEULQCPJDDSLD-UHFFFAOYSA-L
SMILES: [F-].[F-].[Cd++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results