missing translation for 'onlineSavingsMsg'
Learn More

Cadmium fluoride, Puratronic™, 99.99% (metals basis)

Catalog No. AA4020322
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4020322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4020322 Supplier Thermo Scientific Chemicals Supplier No. 04020322
Only null left

CAS: 7790-79-6 | CdF2 | 150.41 g/mol

Cadmium fluoride is used in phosphors, glass and nuclear reactor controls. It is also used in high-temperature lubricants and to start crystals for lasers. It is used in oxygen-sensitive applications such as the production of metallic alloys and it is also used in limited medical treatment protocols. In addition, it can be used in synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7790-79-6
Molecular Formula CdF2
Molecular Weight (g/mol) 150.41
MDL Number MFCD00010919
InChI Key LVEULQCPJDDSLD-UHFFFAOYSA-L
IUPAC Name cadmium(2+) difluoride
SMILES [F-].[F-].[Cd++]

Specifications

Boiling Point 1758°C
Melting Point 1100°C
Physical Form Powder
Quantity 100 g
UN Number UN2570
Merck Index 14,1619
Solubility Information Soluble in water (43.5g/L).
Formula Weight 150.4
Percent Purity 99.99%
Grade Puratronic
Odor Odorless
Density 6.64 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium fluoride
Show More Show Less
Hazard Category H301-H330-H340-H350-H360FD-H372
Hazard Statement GHS H Statement
H301-H330-H340-H350-H360-H372
Toxic if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 232-222-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only