accessibility menu, dialog, popup

UserName

OHN(Z)

CAS RN 779-84-0

IUPAC Name: (6Z)-6-[(phenylamino)methylidene]cyclohexa-2,4-dien-1-one
Synonyms: (6Z)-6-[(phenylamino)methylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 197.24
Molecular Formula: C13H11NO
InChi Key: GVHXCXKLYZOBKS-KHPPLWFESA-N
SMILES: O=C1C=CC=C\C1=C\NC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)