missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Salicylideneaniline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 51°C |
| Boiling Point | 151°C |
| Quantity | 5 g |
| Formula Weight | 197.24 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Salicylideneaniline |
Safety and Handling
| TSCA | No |