accessibility menu, dialog, popup

UserName

CuBrBr

CAS RN 7789-45-9

IUPAC Name: copper(2+) dibromide
Synonyms: copper(II) bromide
Molecular Weight (g/mol): 223.35
Molecular Formula: Br2Cu
InChi Key: QTMDXZNDVAMKGV-UHFFFAOYSA-L
SMILES: [Cu++].[Br-].[Br-]
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
Percent Purity 
  • (4)
  • (6)
  • (5)
Grade 
  • (2)
  • (4)
  • (8)
Physical Form 
  • (2)
  • (4)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (6)
  • (11)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (6)
  • (5)
  • (2)
  • (4)
  • (8)
  • (2)
  • (4)
  • (6)