missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) bromide, 98+%
Description
Copper(II) bromide is used in photographic processing as an intensifier and as a brominating agent in organic synthesis. It is also used in the coupling of o-iodophenols and aryl acetylenes avoiding the use of palladium. It finds application in the preparation of NK1/NK2 receptor antagonists used in the regulation of tachykinin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 900°C |
| Melting Point | 498°C |
| Color | Black |
| Physical Form | Crystalline |
| Quantity | 500 g |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Appearance | Black Crystalline powder |
| Density | 4.71 |
| Solubility Information | Soluble in water,alcohol,ammonia and acetone. Insoluble in benzene,ether,sulfuric acid and ethyl ether. |
| Show More |
Safety and Handling
| Hazard Category | H302-H314 |
| Precautionary Statement | P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | DOT Class: 8, Packaging Group: III |
| EINECSNumber | 232-167-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only