CAS RN 7789-23-3
Potassium Fluoride, Anhydrous (White Powder/Certified), Fisher Chemical™
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 IUPAC Name: potassium fluoride SMILES: [F-].[K+]
Potassium fluoride, anhydrous, 99%, Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 SMILES: [F-].[K+]
Potassium fluoride, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 IUPAC Name: potassium fluoride SMILES: [F-].[K+]
Potassium Fluoride, purum p.a., ≥99.0% (F), Solstice
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 IUPAC Name: potassium;fluoride SMILES: [F-].[K+]
Potassium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 IUPAC Name: potassium fluoride SMILES: [F-].[K+]
Potassium fluoride, ACS, 99% min, Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 SMILES: [F-].[K+]
Potassium fluoride, 99%, extra pure, anhydrous, Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 SMILES: [F-].[K+]
Potassium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 SMILES: [F-].[K+]
Potassium Fluoride, 30% (w/w), Ricca Chemical
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.097 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 IUPAC Name: potassium;fluoride SMILES: [F-].[K+]
Potassium Fluoride, 40% (w/w), Ricca Chemical
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.097 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 IUPAC Name: potassium;fluoride SMILES: [F-].[K+]
Potassium Fluoride, 50% (w/v), Neutralized, Ricca Chemical
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 IUPAC Name: potassium fluoride SMILES: [F-].[K+]
Potassium Fluoride, 40% (w/v), Ricca Chemical
Molecular Formula: FK MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 IUPAC Name: potassium fluoride SMILES: [F-].[K+]
Potassium Fluoride, 50% (w/w), pH 8.3, Ricca Chemical
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 IUPAC Name: potassium fluoride SMILES: [F-].[K+]
Potassium Fluoride, Reagent Grade, LabChem™
CAS: 7789-23-3 Molecular Formula: FK Molecular Weight (g/mol): 58.10 MDL Number: MFCD00011398 InChI Key: NROKBHXJSPEDAR-UHFFFAOYSA-M Synonym: potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568 PubChem CID: 522689 ChEBI: CHEBI:66872 IUPAC Name: potassium fluoride SMILES: [F-].[K+]
- 1
- 2