accessibility menu, dialog, popup

UserName

CH3NBrCl

CAS RN 778611-64-6

IUPAC Name: 5-bromo-2-chloro-4-methylpyridine
Synonyms: 5-bromo-2-chloro-4-methylpyridine
Molecular Weight (g/mol): 206.47
Molecular Formula: C6H5BrClN
InChi Key: ISESOOISZHSENQ-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=NC=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results