missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloro-4-methylpyridine 98.0+%, TCI America™

Catalog No. B46265G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46265G 5 g
B462625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46265G Supplier TCI America Supplier No. B46265G

Chemical Identifiers

CAS 778611-64-6
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD03427663
InChI Key ISESOOISZHSENQ-UHFFFAOYSA-N
Synonym 5-Bromo-2-chloro-4-picoline
PubChem CID 2734427
IUPAC Name 5-bromo-2-chloro-4-methylpyridine
SMILES CC1=CC(=NC=C1Br)Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 206.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Bromo-2-chloro-4-methylpyridine