CAS RN 7784-31-8
Aluminum Sulfate Hydrate (Crystalline/Certified ACS), Fisher Chemical
CAS: 7784-31-8 Molecular Formula: Al2H36O30S3 Molecular Weight (g/mol): 666.401 MDL Number: MFCD00149136 InChI Key: AMVQGJHFDJVOOB-UHFFFAOYSA-H Synonym: aluminum sulfate octadecahydrate,patent alum,cake alum octadecahydrate,unii-tcs9l00g8f,dialuminum sulfate octadecahydrate,tcs9l00g8f,aluminum sulfate.18h2o,aluminum sulfate.18h2o,dialuminium sulfate octadecahydrate,aluminum sulfate-water 1/18 PubChem CID: 22377415 ChEBI: CHEBI:74779 IUPAC Name: dialuminum;trisulfate;octadecahydrate SMILES: O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
Aluminum Sulfate, 18-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 7784-31-8 Molecular Formula: Al2H36O30S3 Molecular Weight (g/mol): 666.401 InChI Key: AMVQGJHFDJVOOB-UHFFFAOYSA-H Synonym: aluminum sulfate octadecahydrate,patent alum,cake alum octadecahydrate,unii-tcs9l00g8f,dialuminum sulfate octadecahydrate,tcs9l00g8f,aluminum sulfate.18h2o,aluminum sulfate.18h2o,dialuminium sulfate octadecahydrate,aluminum sulfate-water 1/18 PubChem CID: 22377415 ChEBI: CHEBI:74779 IUPAC Name: dialuminum;trisulfate;octadecahydrate SMILES: O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
Aluminum Sulfate ACS AR Crystal, Macron Fine Chemicals™
CAS: 7784-31-8 Molecular Formula: Al2H36O30S3 Molecular Weight (g/mol): 666.401 InChI Key: AMVQGJHFDJVOOB-UHFFFAOYSA-H Synonym: aluminum sulfate octadecahydrate,patent alum,cake alum octadecahydrate,unii-tcs9l00g8f,dialuminum sulfate octadecahydrate,tcs9l00g8f,aluminum sulfate.18h2o,aluminum sulfate.18h2o,dialuminium sulfate octadecahydrate,aluminum sulfate-water 1/18 PubChem CID: 22377415 ChEBI: CHEBI:74779 IUPAC Name: dialuminum;trisulfate;octadecahydrate SMILES: O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
Aluminum sulfate octadecahydrate, 98+%, ACS reagent
CAS: 7784-31-8 Molecular Formula: Al2O12S3·18H2O Molecular Weight (g/mol): 666.4 MDL Number: MFCD00149136 InChI Key: AMVQGJHFDJVOOB-UHFFFAOYSA-H Synonym: aluminum sulfate octadecahydrate,patent alum,cake alum octadecahydrate,unii-tcs9l00g8f,dialuminum sulfate octadecahydrate,tcs9l00g8f,aluminum sulfate.18h2o,aluminum sulfate.18h2o,dialuminium sulfate octadecahydrate,aluminum sulfate-water 1/18 PubChem CID: 22377415 ChEBI: CHEBI:74779 IUPAC Name: dialuminum;trisulfate;octadecahydrate SMILES: O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
Aluminum sulfate octadecahydrate, extra pure
CAS: 7784-31-8 Molecular Formula: Al2O12S3·18H2O Molecular Weight (g/mol): 666.4 MDL Number: MFCD00149136 InChI Key: AMVQGJHFDJVOOB-UHFFFAOYSA-H Synonym: aluminum sulfate octadecahydrate,patent alum,cake alum octadecahydrate,unii-tcs9l00g8f,dialuminum sulfate octadecahydrate,tcs9l00g8f,aluminum sulfate.18h2o,aluminum sulfate.18h2o,dialuminium sulfate octadecahydrate,aluminum sulfate-water 1/18 PubChem CID: 22377415 ChEBI: CHEBI:74779 IUPAC Name: dialuminum;trisulfate;octadecahydrate SMILES: O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
Aluminum Sulfate, 20% (w/w), Ricca Chemical
CAS: 7784-31-8 Molecular Formula: Al2H36O30S3 Molecular Weight (g/mol): 666.401 InChI Key: AMVQGJHFDJVOOB-UHFFFAOYSA-H Synonym: aluminum sulfate octadecahydrate,patent alum,cake alum octadecahydrate,unii-tcs9l00g8f,dialuminum sulfate octadecahydrate,tcs9l00g8f,aluminum sulfate.18h2o,aluminium sulfate.18h2o,dialuminium sulfate octadecahydrate,aluminium sulfate-water 1/18 PubChem CID: 22377415 ChEBI: CHEBI:74779 IUPAC Name: dialuminum;trisulfate;octadecahydrate SMILES: O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]
Aluminum Sulfate, Hydrated, Crystal, ACS, 98-102%, Spectrum™ Chemical
CAS: 7784-31-8
Aluminum Sulfate, Crystal, FCC, 99.5-114%, Spectrum™ Chemical
Aluminum sulfate hydrate, Puriss., meets analytical spec. of BP, Ph. Eur., 100-110%, 51.0-59.0% Al2(SO4)3 basis, Solstice
CAS: 7784-31-8 Molecular Formula: Al2H36O30S3 Molecular Weight (g/mol): 666.401 InChI Key: AMVQGJHFDJVOOB-UHFFFAOYSA-H Synonym: aluminum sulfate octadecahydrate,patent alum,cake alum octadecahydrate,unii-tcs9l00g8f,dialuminum sulfate octadecahydrate,tcs9l00g8f,aluminum sulfate.18h2o,aluminum sulfate.18h2o,dialuminium sulfate octadecahydrate,aluminum sulfate-water 1/18 PubChem CID: 22377415 ChEBI: CHEBI:74779 IUPAC Name: dialuminum;trisulfate;octadecahydrate SMILES: O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]