missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sulfate hydrate, Puriss., meets analytical spec. of BP, Ph. Eur., 100-110%, 51.0-59.0% Al2(SO4)3 basis, Solstice

Catalog No. 60044687
Change view
Click to view available options
Quantity:
6 x 1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60044687 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60044687 Supplier Solstice Supplier No. 110446X1KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss., meets analytical specification of BP, Ph. Eur., 100-110%, 51.0-59.0% Al2(SO4)3 basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7784-31-8
Molecular Formula Al2H36O30S3
Molecular Weight (g/mol) 666.401
InChI Key AMVQGJHFDJVOOB-UHFFFAOYSA-H
Synonym aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
PubChem CID 22377415
ChEBI CHEBI:74779
IUPAC Name dialuminum;trisulfate;octadecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Specifications

Quantity 6 x 1 kg
Linear Formula Al2(SO4)3 · 18H2O
UN Number UN3260
Formula Weight 342.15 (anhydrous basis)
Percent Purity 100 to 110%
Grade puriss.
Chemical Name or Material Aluminum Sulfate Hydrate, Meets analytical specification of Ph.Eur.BP
Hazard Statement P280-P305 + P351 + P338 + P310
Precautionary Statement H290-H318
DOTInformation Class 8 - PG 3 - RQ - Corrosive solid, acidic, inorganic, n.o.s., HI: all
EINECSNumber 233-135-0
RTECSNumber WS5697000