missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Sulfate, Hydrated, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18608555
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608555 50 kg Poly Bottle
18608551 125 g Poly Bottle
18608552 500 g Poly Bottle
18608553 2.5 kg Poly Bottle
18608554 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608555 Supplier Spectrum Chemical Mfg Cor Supplier No. A111550KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7784-31-8

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Molecular Weight (g/mol) 342.14
Percent Purity 98 to 102%
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Aluminum Sulfate, Hydrated

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.