missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Sulfate, 18-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Specifications
Specifications
| Physical Form | Crystals |
| Quantity | 500 g |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 1.69kg |
| Formula Weight | 666.41 |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Chemical Name or Material | Aluminum Sulfate, 18-Hydrate |