missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Sulfate, 20% (w/w), Ricca Chemical

Catalog No. R06075004A
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R06075004A 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R06075004A Supplier Ricca Chemical Company Supplier No. R06075004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7784-31-8 , 7732-18-5
Molecular Formula Al2H36O30S3
Molecular Weight (g/mol) 666.401
InChI Key AMVQGJHFDJVOOB-UHFFFAOYSA-H
Synonym aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminium sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminium sulfate-water 1/18
PubChem CID 22377415
ChEBI CHEBI:74779
IUPAC Name dialuminum;trisulfate;octadecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Specifications

Quantity 4 L
Assay Percent Range 20% (w/w)
Odor odorless
Packaging Natural Poly Bottle
Chemical Name or Material Aluminum Sulfate
ShelfLife 24 months