accessibility menu, dialog, popup

UserName

CAS RN 7766-50-9

IUPAC Name: 11-bromoundec-1-ene
Synonyms: 11-bromoundec-1-ene
Molecular Weight (g/mol): 233.19
Molecular Formula: C11H21Br
InChi Key: YPLVPFUSXYSHJD-UHFFFAOYSA-N
SMILES: BrCCCCCCCCCC=C
Boiling Point 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)