missing translation for 'onlineSavingsMsg'
Learn More

11-Bromo-1-undecene 94.0+%, TCI America™

Catalog No. B35765G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35765G 5 g
B357625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35765G Supplier TCI America Supplier No. B35765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7766-50-9
Molecular Formula C11H21Br
Molecular Weight (g/mol) 233.193
MDL Number MFCD00040825
InChI Key YPLVPFUSXYSHJD-UHFFFAOYSA-N
Synonym 11-bromo-1-undecene, 10-undecenyl bromide, 1-bromo-10-undecene, undec-10-enyl bromide, 11-bromo-undec-1-ene, 1-undecene, 11-bromo, 11-bromoundecene, undecylenyl bromide, n-undecylenic bromide, w-undecylenyl bromide
PubChem CID 284148
IUPAC Name 11-bromoundec-1-ene
SMILES C=CCCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 115°C
Quantity 5 g
Formula Weight 233.19
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 11-Bromo-1-undecene
EINECSNumber (2)-0067
TSCA No