missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate Solution (1N/Certified), Fisher Chemical

Catalog No. SS68500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SS68500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SS68500 Supplier Thermo Fisher Scientific Supplier No. FLSS68500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Standardized at 25°C against NIST™ potassium chloride

Chemical Identifiers

CAS 7761-88-8 , 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
UN Number 1493
Concentration 0.995 to 1.005N
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 14mmHg at 20°C
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Silver Nitrate Solution, 1N