missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate (Cryst./Molecular Biology Grade), Fisher BioReagents

Catalog No. BP2546100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP2546100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP2546100 Supplier Fisher BioReagents Supplier No. BP2546100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For silver staining

Silver Nitrate is used in biochemistry to stain agarose and acrylamide gels. In histology, silver nitrate is used for silver staining to demonstrate reticular fibers, proteins and nucleic acids.

Specifications

Boiling Point 444°C
Melting Point 212°C
Color White
CAS 7761-88-8
pH 5.4 to 6.4
Physical Form Solid
Quantity 100 g
Assay Percent Range ≥99.7 %
Molecular Formula AgNO3
MDL Number MFCD00003414
Merck Index 15, 8657
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES [Ag+].[O-][N+]([O-])=O
IUPAC Name silver(1+) nitrate
Molecular Weight (g/mol) 169.87
PubChem CID 24470
ChEBI CHEBI:32130
Formula Weight 169.87
Percent Purity ≥99.7%
Grade Molecular Biology
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Silver Nitrate
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 5.1, : Oxidizer
Recommended Storage RT