accessibility menu, dialog, popup

UserName

HOOHN

CAS RN 771-50-6

IUPAC Name: 1H-indole-3-carboxylic acid
Synonyms: indole - 3 carboxylic acid indole-3-carboxylicacid
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: KMAKOBLIOCQGJP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CNC2=CC=CC=C12
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)