accessibility menu, dialog, popup

UserName

CAS RN 77-57-6

IUPAC Name: 1-phenylcyclopentane-1-carbonitrile
Synonyms: 1-phenylcyclopentane-1-carbonitrile
Molecular Weight (g/mol): 171.24
Molecular Formula: C12H13N
InChi Key: GDXMFFGTPGAGGX-UHFFFAOYSA-N
SMILES: N#CC1(CCCC1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results