CAS RN 7695-91-2
Thermo Scientific Chemicals DL-alpha-Tocopheryl acetate, 98%
CAS: 7695-91-2 Molecular Formula: C31H52O3 Molecular Weight (g/mol): 472.75 MDL Number: MFCD00072042 InChI Key: ZAKOWWREFLAJOT-CEFNRUSXSA-N Synonym: vitamin e acetate,tocopherol acetate,alpha-tocopherol acetate,alfacol,ecofrol,d-alpha-tocopherol acetate,contopheron,tofaxin,ephynal acetate,econ PubChem CID: 86472 ChEBI: CHEBI:32321 IUPAC Name: [(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate SMILES: CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C
Vitamin E acetate, 97%
CAS: 7695-91-2 Molecular Formula: C31H52O3 Molecular Weight (g/mol): 472.754 MDL Number: MFCD00072042 InChI Key: ZAKOWWREFLAJOT-CEFNRUSXSA-N Synonym: vitamin e acetate,tocopherol acetate,alpha-tocopherol acetate,alfacol,ecofrol,d-alpha-tocopherol acetate,contopheron,tofaxin,ephynal acetate,econ PubChem CID: 86472 ChEBI: CHEBI:32321 IUPAC Name: [(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate SMILES: CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C
Tocopheryl Acetate, a Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards
DL-alpha-Tocopherol Acetate 97.0+%, TCI America™
CAS: 7695-91-2 Molecular Formula: C31H52O3 Molecular Weight (g/mol): 472.754 MDL Number: MFCD00072042 InChI Key: ZAKOWWREFLAJOT-CEFNRUSXSA-N Synonym: vitamin e acetate,tocopherol acetate,alpha-tocopherol acetate,alfacol,ecofrol,d-alpha-tocopherol acetate,contopheron,tofaxin,ephynal acetate,econ PubChem CID: 86472 ChEBI: CHEBI:32321 IUPAC Name: [(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate SMILES: CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C
All-rac-a-Tocopheryl Acetate, FCC, 96-102%, Spectrum™ Chemical
CAS: 7695-91-2 Molecular Formula: C31H52O3 Molecular Weight (g/mol): 472.75 InChI Key: ZAKOWWREFLAJOT-CEFNRUSXSA-N IUPAC Name: (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate SMILES: CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1
Vitamin E Acetate, USP, 96-102%, Spectrum™ Chemical
CAS: 7695-91-2 Molecular Formula: C31H52O3 Molecular Weight (g/mol): 472.75 InChI Key: ZAKOWWREFLAJOT-CEFNRUSXSA-N IUPAC Name: (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate SMILES: CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1
Vitamin E Acetate, USP, 96-102%, Spectrum™ Chemical
CAS: 7695-91-2 Molecular Formula: C31H52O3 Molecular Weight (g/mol): 472.75 InChI Key: ZAKOWWREFLAJOT-CEFNRUSXSA-N IUPAC Name: (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate SMILES: CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1