missing translation for 'onlineSavingsMsg'
Learn More

Vitamin E Acetate, USP, 96-102%, Spectrum™ Chemical

Catalog No. 18607724
Change view
Click to view available options
Quantity:
20 kg
Packaging:
Steel Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607724 20 kg Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18607724 Supplier Spectrum Chemical Mfg Cor Supplier No. VI14020KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

VI140, 7695-91-2, C31H52O3

Spectrum™ Chemical Vitamin E Acetate, USP is used as an antioxidant. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7695-91-2
Molecular Formula C31H52O3
Molecular Weight (g/mol) 472.75
InChI Key ZAKOWWREFLAJOT-CEFNRUSXSA-N
IUPAC Name (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1

Specifications

Acidity 1.0 mL
CAS Max % 1
Packaging Steel Pail
Quantity 20 kg
Percent Purity 96 to 102%
Grade USP