missing translation for 'onlineSavingsMsg'
Learn More

Tocopheryl Acetate, a Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015480
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015480 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015480 Supplier Merck Emd Millipore Supplier No. PHR1030500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards

Specifications

CAS 7695-91-2
Density 0.96 g/mL (at 20°C)
Flash Point 225.5°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C31H52O3
MDL Number MFCD00072042
Synonym DL-alpha-Tocopherol acetate; Vitamin E acetate; all-rac-alpha-Tocopheryl acetate
Type Certified Reference Material
Quantity 500 mg
Formula Weight 472.74
Grade Certified Reference Material
Show More Show Less