accessibility menu, dialog, popup

UserName

CAS RN 76823-03-5

IUPAC Name: 3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid
Synonyms: 5(6)-carboxyfluorescein
Molecular Weight (g/mol): 376.32
Molecular Formula: C21H12O7
InChi Key: NJYVEMPWNAYQQN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C(=C1)C(=O)OC21C2=CC=C(O)C=C2OC2=CC(O)=CC=C12
Percent Purity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)