missing translation for 'onlineSavingsMsg'
Learn More

5-Carboxyfluorescein Hydrate 97.0+%, TCI America™

Catalog No. C2477100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2477100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2477100MG Supplier TCI America Supplier No. C2477100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76823-03-5
Molecular Formula C21H12O7
Molecular Weight (g/mol) 376.32
MDL Number MFCD00036874
InChI Key NJYVEMPWNAYQQN-UHFFFAOYSA-N
Synonym 5-carboxyfluorescein, 5-fam, 4-carboxyfluorescein, 4 5-carboxyfluorescein, 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-5-carboxylic acid, 3',6'-dihydroxy-3-oxo-3h-spiro 2-benzofuran-1,9'-xanthene-5-carboxylic acid, 3',6'-dihydroxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5-carboxylic acid, spiro isobenzofuran-1 3h ,9'-9h xanthene-5-carboxylic acid, 3',6-dihydroxy-3-oxo, bidd:gt0758, 5-carboxy fluorescein; 5-fam
PubChem CID 123755
ChEBI CHEBI:51617
IUPAC Name 3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid
SMILES OC(=O)C1=CC=C2C(=C1)C(=O)OC21C2=CC=C(O)C=C2OC2=CC(O)=CC=C12

Specifications

Quantity 100 mg
Formula Weight 376.32
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Carboxyfluorescein Hydrate