accessibility menu, dialog, popup

UserName

ClCH3N

CAS RN 7680-73-1

IUPAC Name: 1-methylpyridin-1-ium chloride
Synonyms: methylpyridinium
Molecular Weight (g/mol): 129.59
Molecular Formula: C6H8ClN
InChi Key: QAIGYXWRIHZZAA-UHFFFAOYSA-M
SMILES: [Cl-].C[N+]1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results