missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyridinium Chloride 98.0+%, TCI America™

Catalog No. M13975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13975G 5 g
M139725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13975G Supplier TCI America Supplier No. M13975G

Chemical Identifiers

CAS 7680-73-1
Molecular Formula C6H8ClN
Molecular Weight (g/mol) 129.59
MDL Number MFCD00137532
InChI Key QAIGYXWRIHZZAA-UHFFFAOYSA-M
PubChem CID 82116
IUPAC Name 1-methylpyridin-1-ium chloride
SMILES [Cl-].C[N+]1=CC=CC=C1

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 129.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylpyridinium Chloride
RTECSNumber UU5950000
TSCA No