accessibility menu, dialog, popup

UserName

ClN

CAS RN 766-84-7

IUPAC Name: 3-chlorobenzonitrile
Synonyms: 3-chlorobenzonitrile
Molecular Weight (g/mol): 137.57
Molecular Formula: C7H4ClN
InChi Key: WBUOVKBZJOIOAE-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)C#N
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)