accessibility menu, dialog, popup

UserName

CH3OCH3O(E)

CAS RN 76238-22-7

IUPAC Name: (6E)-non-6-en-1-yl acetate
Synonyms: cis-6-nonenyl acetate
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: DFRXMRZQBMFKMI-SNAWJCMRSA-N
SMILES: CC\C=C\CCCCCOC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results