missing translation for 'onlineSavingsMsg'
Learn More

cis-6-Nonenyl Acetate 92.0+%, TCI America™

Catalog No. A09215ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09215ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A09215ML Supplier TCI America Supplier No. A09215ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76238-22-7
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.28
MDL Number MFCD00059409
InChI Key DFRXMRZQBMFKMI-SNAWJCMRSA-N
Synonym Acetic Acid cis-6-Nonenyl Ester
PubChem CID 5363389
IUPAC Name (6E)-non-6-en-1-yl acetate
SMILES CC\C=C\CCCCCOC(C)=O

Specifications

Color Colorless
Quantity 5 mL
Formula Weight 184.28
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-6-Nonenyl Acetate
EINECSNumber (2)-0762
TSCA Yes