accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3COH

CAS RN 760989-91-1

IUPAC Name: 4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-ol
Synonyms: 4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-ol
Molecular Weight (g/mol): 296.17
Molecular Formula: C18H21BO3
InChi Key: BFQSVDDJLRMXOO-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results