accessibility menu, dialog, popup

UserName

CH3H3COBOH3CH3CCH2

CAS RN 75927-49-0

IUPAC Name: 2-ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 154.02
Molecular Formula: C8H15BO2
InChi Key: DPGSPRJLAZGUBQ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C=C
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)