missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane (stabilized with Phenothiazine) 93.0+%, TCI America™

Catalog No. T22975G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22975G 5 g
T22971G 1 g
T229725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T22975G Supplier TCI America Supplier No. T22975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75927-49-0
Molecular Formula C8H15BO2
Molecular Weight (g/mol) 154.02
MDL Number MFCD00192492
InChI Key DPGSPRJLAZGUBQ-UHFFFAOYSA-N
Synonym vinylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-vinyl-1,3,2-dioxaborolane, vinylboronic acid pinacol cyclic ester, 2-vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-ethenyl-4,4,5,5-tetramethyl, pinacol vinylboronate, stabilized with bht, 2-vinyl-4,4,5,5-tetramethyl-1,3,2-dioxaoborolane, vbpe, pubchem23743, ethenylboronic acid pinacol ester
PubChem CID 5233012
IUPAC Name 2-ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C=C

Specifications

Color Yellow
Boiling Point 40°C
Quantity 5 g
UN Number 3272
Formula Weight 154.02
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane (stabilized with Phenothiazine)
TSCA No
Recommended Storage Refrigerator