accessibility menu, dialog, popup

UserName

H3COONNCl

CAS RN 75680-92-1

IUPAC Name: ethyl 6-chloropyridazine-3-carboxylate
Synonyms: ethyl 6-chloropyridazine-3-carboxylate
Molecular Weight (g/mol): 186.6
Molecular Formula: C7H7ClN2O2
InChi Key: GVSVPKDEHFOXSW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NN=C(Cl)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)