missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-chloropyridazine-3-carboxylate, 95%

Catalog No. AC431110050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431110050 5 g Glass bottle
AC431110010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC431110050 Supplier Thermo Scientific Chemicals Supplier No. 431110050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75680-92-1 | C7H7ClN2O2 | 186.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75680-92-1
Molecular Formula C7H7ClN2O2
Molecular Weight (g/mol) 186.6
MDL Number MFCD09908168
InChI Key GVSVPKDEHFOXSW-UHFFFAOYSA-N
Synonym ethyl 6-chloro-3-pyridazinecarboxylate, 6-chloro-pyridazine-3-carboxylic acid ethyl ester, 3-pyridazinecarboxylic acid, 6-chloro-, ethyl ester, 6-chloropyridazine-3-carboxylic acid ethyl ester, 6-chloro-3-pyridazinecarboxylic acid ethyl ester, pubchem17694, acmc-1bd9t, 6-chloro-pyridazine-3-carboxylicacidethylester, ksc641q6n, ethyl 3-chloropyridazine-6-carboxylate
PubChem CID 10352425
IUPAC Name ethyl 6-chloropyridazine-3-carboxylate
SMILES CCOC(=O)C1=NN=C(C=C1)Cl

Specifications

Melting Point 149.0°C to 152.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 186.6
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 6-chloropyridazine-3-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only